L-Argininamide dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Arg-NH₂·2HCl, H-Arg-NH₂·2 HCl, H-Arg-NH2.2HCl

A152216-1G A152216-100G A152216-25G A152216-5G
76997-386EA 10 USD
76997-386 77600-160 77600-162 77600-164
L-Argininamide dihydrochloride 98%
L-Argininamide dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₇Cl₂N₅O
MDL Number: MFCD00058286
CAS Number: 14975-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR