2-Chloro-4-methoxypyrimidin-5-amine 97%

Supplier: Ambeed
Warning

A152007-100MG A152007-1G A152007-250MG
77010-842EA 42.44 USD
77010-842 77010-844 77010-846
2-Chloro-4-methoxypyrimidin-5-amine 97%
2-Chloro-4-methoxypyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃O
MW: 159.57 g/mol
MDL Number: MFCD12402341
CAS Number: 96833-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR