6-(Trifluoromethyl)-1-indanone 95%

Supplier: Ambeed
Warning

A151987-100MG A151987-10G A151987-250MG A151987-25G A151987-5G A151987-1G
77028-178EA 6 USD
77028-178 77028-180 77028-182 77028-184 77592-856 77667-152
6-(Trifluoromethyl)-1-indanone 95%
6-(Trifluoromethyl)-1-indanone

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O
MW: 200.157 g/mol
MDL Number: MFCD07772123
CAS Number: 68755-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR