(S)-3-[1-(Dimethylamino)ethyl]phenol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-3-(1-(Dimethylamino)ethyl)phenol, NAP-226-90
A151961-100G
A151961-1G
A151961-250MG
A151961-25G
A151961-5G
A151961-10G
76997-376EA
751.12
USD
76997-376
76997-378
76997-380
76997-382
76997-384
77671-130
(S)-3-[1-(Dimethylamino)ethyl]phenol 98%
(S)-3-[1-(Dimethylamino)ethyl]phenol
Precaución: for Research Use Only
Formula:
C₁₀H₁₅NO MW: 165.24 g/mol |
MDL Number:
MFCD06656491 CAS Number: 139306-10-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...