Bicyclo[2.2.2]octane-1,4-dicarboxylic acid 96%

Supplier: Ambeed
Warning

A151946-1G A151946-25G A151946-5G A151946-100G
77028-168EA 16 USD
77028-168 77028-170 77028-172 77871-750
Bicyclo[2.2.2]octane-1,4-dicarboxylic acid 96%
Bicyclo[2.2.2]octane-1,4-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄
MDL Number: MFCD00498000
CAS Number: 711-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR