4-Methylpyrazolo[1,5-a]pyridine-3-carboxylic acid 96%

Supplier: Ambeed
Warning

A151884-100MG A151884-250MG
77010-838EA 193.09 USD
77010-838 77010-840
4-Methylpyrazolo[1,5-a]pyridine-3-carboxylic acid 96%
4-Methylpyrazolo[1,5-a]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
MDL Number: MFCD12405114
CAS Number: 143803-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR