5-Fluoro-3-nitropyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-hydroxy-3-nitropyridine

A151859-100G A151859-100MG A151859-1G A151859-250MG A151859-25G A151859-5G
76997-362EA 1122.44 USD
76997-362 76997-364 76997-366 76997-368 76997-370 76997-372
5-Fluoro-3-nitropyridin-2(1H)-one 95%
5-Fluoro-3-nitropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C5H3FN2O3
MW: 158.09 g/mol
MDL Number: MFCD05662412
CAS Number: 136888-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR