Boc-D-aspartic acid β-benzyl ester 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-D-aspartic acid 4-benzyl ester, Boc-D-Asp(OBzl)-OH

A151838-100G A151838-10G A151838-1G A151838-250MG A151838-25G A151838-500G A151838-5G
77028-148EA 115 USD
77028-148 77028-150 77028-152 77028-154 77028-156 77028-158 77028-160
Boc-D-aspartic acid β-benzyl ester 97%
Boc-D-aspartic acid beta-benzyl ester

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₆
MW: 323.345 g/mol
MDL Number: MFCD00038255
CAS Number: 51186-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR