1,3,5-Tris(4-aminophenyl)benzene 97%

Supplier: Ambeed
Warning

A151591-100MG A151591-1G A151591-250MG A151591-25G A151591-5G A151591-100G
76984-654EA 7 USD
76984-654 76984-656 76984-658 76984-660 76984-662 77889-997
1,3,5-Tris(4-aminophenyl)benzene 97%
1,3,5-Tris(4-aminophenyl)benzene

Precaución: for Research Use Only

Formula: C₂₄H₂₁N₃
MW: 351.45 g/mol
MDL Number: MFCD18207723
CAS Number: 118727-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR