2,5-Dibromopyridin-4-amine 95%

Supplier: Ambeed
Warning

A151435-100MG A151435-1G A151435-250MG
77010-812EA 26.53 USD
77010-812 77010-814 77010-816
2,5-Dibromopyridin-4-amine 95%
2,5-Dibromopyridin-4-amine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MW: 251.90 g/mol
MDL Number: MFCD13185831
CAS Number: 221241-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR