2-(2-Hydroxyethoxy)ethyl 4-methylbenzenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Tos-PEG2-OH

A151206-100MG A151206-250MG A151206-1G
77010-786EA 17 USD
77010-786 77010-788 77889-992
2-(2-Hydroxyethoxy)ethyl 4-methylbenzenesulfonate 98%
2-(2-Hydroxyethoxy)ethyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₅S
MDL Number: MFCD22574797
CAS Number: 118591-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR