1-(3-Aminopyridin-4-yl)ethanone 95%

Supplier: Ambeed
Warning

Synonyms: 4-Acetyl-3-aminopyridine

A151205-100MG A151205-10G A151205-1G A151205-250MG A151205-5G
77010-776EA 19.1 USD
77010-776 77010-778 77010-780 77010-782 77010-784
1-(3-Aminopyridin-4-yl)ethanone 95%
1-(3-Aminopyridin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD07375078
CAS Number: 13210-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR