Methyl-5-methyl-2-nitrobenzoate 98%
Supplier: Ambeed
|
Warning
|
A151183-100G
A151183-10G
A151183-1G
A151183-25G
A151183-500G
A151183-5G
77010-764EA
43
USD
77010-764
77010-766
77010-768
77010-770
77010-772
77010-774
Methyl-5-methyl-2-nitrobenzoate 98%
Methyl 5-methyl-2-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₉H₉NO₄ |
MDL Number:
MFCD00157275 CAS Number: 20587-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...