5'-Fluoro-2'-iodoacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Fluoro-2-iodophenyl)ethanone

A151174-1G A151174-25G A151174-5G
77028-130EA 6.37 USD
77028-130 77028-132 77028-134
5'-Fluoro-2'-iodoacetophenone 98%
5'-Fluoro-2'-iodoacetophenone

Precaución: for Research Use Only

Formula: C₈H₆FIO
MW: 264.038 g/mol
MDL Number: MFCD09033159
CAS Number: 914225-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR