N-(Piperidin-4-ylmethyl)methanesulfonamide hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A151125-100MG
A151125-1G
A151125-250MG
77028-124EA
48
USD
77028-124
77028-126
77028-128
N-(Piperidin-4-ylmethyl)methanesulfonamide hydrochloride 95%
N-(Piperidin-4-ylmethyl)methanesulfonamide hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₇ClN₂O₂S |
MDL Number:
MFCD08461902 CAS Number: 1185092-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...