[3-(Trifluoromethoxy)phenyl]hydrazine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (3-(Trifluoromethoxy)phenyl)hydrazine hydrochloride

A151029-1G A151029-5G A151029-25G
77010-754EA 11 USD
77010-754 77010-756 77010-758 77889-991
[3-(Trifluoromethoxy)phenyl]hydrazine hydrochloride 97%
[3-(Trifluoromethoxy)phenyl]hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇F₃N₂O·ClH
MW: 228.599 g/mol
MDL Number: MFCD02671043
CAS Number: 133115-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR