(R)-Ethyl 3-methylpiperidine-3-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A150990-1G
A150990-250MG
77010-752EA
155
USD
77010-752
77889-990
(R)-Ethyl 3-methylpiperidine-3-carboxylate 95%
(R)-Ethyl 3-methylpiperidine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₂ |
MDL Number:
MFCD13181584 CAS Number: 297172-01-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...