Neratinib 98%

Supplier: Ambeed
Warning

Synonyms: HKI-272, (E)-N-(4-((3-Chloro-4-(pyridin-2-ylmethoxy)phenyl)amino)-3-cyano-7-ethoxyquinolin-6-yl)-4-(dimethylamino)but-2-enamide

A150970-10MG A150970-1MG A150970-250MG A150970-100MG A150970-1G A150970-5MG A150970-5G
77028-118EA 8 USD
77028-118 77028-120 77028-122 77600-138 77600-140 77600-142 77667-136
Neratinib 98%
Neratinib

Precaución: for Research Use Only

Formula: C₃₀H₂₉ClN₆O₃
MW: 557 g/mol
MDL Number: MFCD09752958
CAS Number: 698387-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR