LY-2157299 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4-(2-(6-Methylpyridin-2-yl)-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazol-3-yl)quinoline-6-carboxamide, Galunisertib

A150887-10MG A150887-25MG A150887-5MG A150887-1G A150887-1MG A150887-2MG A150887-100MG
77010-746EA 18 USD
77010-746 77010-748 77010-750 77611-402 77611-404 77611-406 77683-274
LY-2157299 98%, Ambeed.Inc
LY-2157299

Precaución: For research use only.

Formula: C₂₂H₁₉N₅O
MW: 369.42 g/mol
MDL Number: MFCD12923319
CAS Number: 700874-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR