2-Amino-6-chloro-4-fluorotoluene 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-5-fluoro-2-methylaniline

A150861-1G A150861-250MG
76984-644EA 242.95 USD
76984-644 76984-646
2-Amino-6-chloro-4-fluorotoluene 95%
2-Amino-6-chloro-4-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₇ClFN
MW: 159.589 g/mol
MDL Number: MFCD03094195
CAS Number: 886761-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR