Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate 95%

Supplier: Ambeed
Warning

A150816-1G A150816-250MG A150816-5G
76997-330EA 12 USD
76997-330 76997-332 77671-122
Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate 95%
Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MDL Number: MFCD00847745
CAS Number: 115932-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR