2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile 95%

Supplier: Ambeed
Warning

A150676-1G A150676-25G A150676-5G
77028-108EA 12 USD
77028-108 77028-110 77028-112 77028-114
2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile 95%
2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₅ClF₃N
MDL Number: MFCD04115850
CAS Number: 886496-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR