3-Ethoxy-5-fluorobenzeneboronic acid 97%

Supplier: Ambeed
Warning

A150640-1G A150640-250MG
77028-104EA 46 USD
77028-104 77871-737
3-Ethoxy-5-fluorobenzeneboronic acid 97%
3-Ethoxy-5-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MDL Number: MFCD07363779
CAS Number: 850589-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR