3-Chloro-2-formylbenzonitrile 98%

Supplier: Ambeed
Warning

A150623-100MG A150623-1G A150623-250MG
77010-734EA 197 USD
77010-734 77010-736 77010-738
3-Chloro-2-formylbenzonitrile 98%
3-Chloro-2-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClNO
MW: 165.576 g/mol
MDL Number: MFCD16251143
CAS Number: 1256561-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR