6-Chloro-2-fluoronicotinic acid 97%

Supplier: Ambeed
Warning

A150547-100G A150547-1G A150547-250MG A150547-25G A150547-5G
76984-624EA 2673 USD
76984-624 76984-626 76984-628 76984-630 76984-632
6-Chloro-2-fluoronicotinic acid 97%
6-Chloro-2-fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MDL Number: MFCD16610397
CAS Number: 1211578-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR