N-Methoxy-N,1-dimethylpiperidine-4-carboxamide 95%

Supplier: Ambeed
Warning

A150526-1G A150526-250MG A150526-25G A150526-5G
76984-616EA 28.65 USD
76984-616 76984-618 76984-620 76984-622
N-Methoxy-N,1-dimethylpiperidine-4-carboxamide 95%
N-Methoxy-N,1-dimethylpiperidine-4-carboxamide

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MW: 186.251 g/mol
MDL Number: MFCD22575146
CAS Number: 215950-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR