Ibrutinib 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-1-(3-(4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidin-1-yl)prop-2-en-1-one, PCI-32765
A150396-10G
A150396-10MG
A150396-250MG
A150396-1G
A150396-100MG
A150396-5G
A150396-5MG
76984-604EA
332.07
USD
76984-604
76984-606
76984-608
77549-886
77611-394
77611-396
77683-268
Ibrutinib 98%
Ibrutinib
Precaución: For research use only.
Formula:
C₂₅H₂₄N₆O₂ MW: 440.5 g/mol |
MDL Number:
MFCD20261150 CAS Number: 936563-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...