Ethyl-4-fluoroindole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-fluoro-1H-indole-2-carboxylate

A150388-100MG A150388-1G A150388-250MG A150388-5G A150388-10G
77028-096EA 8 USD
77028-096 77028-098 77028-100 77028-102 77871-736
Ethyl-4-fluoroindole-2-carboxylate 97%
Ethyl-4-fluoroindole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀FNO₂
MDL Number: MFCD09264533
CAS Number: 348-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR