Ethyl 5-bromo-3-methylpicolinate 99%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-methylpyridine-2-carboxylic acid ethyl ester

A150171-100G A150171-10G A150171-1G A150171-25G A150171-5G
76984-586EA 571.83 USD
76984-586 76984-588 76984-590 76984-592 76984-594
Ethyl 5-bromo-3-methylpicolinate 99%
Ethyl 5-bromo-3-methylpicolinate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.09 g/mol
MDL Number: MFCD15523779
CAS Number: 794592-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR