5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine 97%
Supplier: Ambeed
|
Warning
|
A149976-100MG
A149976-1G
A149976-250MG
A149976-5G
77010-706EA
24
USD
77010-706
77010-708
77010-710
77567-144
5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine 97%
5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine
Precaución: For research use only.
Formula:
C₆H₅BrFNO |
MDL Number:
MFCD16607004 CAS Number: 1206968-92-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...