1-Methyl-1H-indazole-6-carboxylic acid methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-1H-indazole-6-carboxylate

A149815-1G A149815-250MG A149815-5G
77028-074EA 42.44 USD
77028-074 77028-076 77028-078
1-Methyl-1H-indazole-6-carboxylic acid methyl ester 97%
1-Methyl-1H-indazole-6-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
MDL Number: MFCD11109403
CAS Number: 1007219-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR