1-Ethyl-5-nitroindazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Ethyl-5-nitro-1H-indazole

A149653-100MG A149653-1G A149653-250MG
77010-696EA 18 USD
77010-696 77010-698 77010-700
1-Ethyl-5-nitroindazole 98%
1-Ethyl-5-nitroindazole

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MDL Number: MFCD20491412
CAS Number: 5228-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR