3-Amino-3-(3-methoxyphenyl)propan-1-ol 95%

Supplier: Ambeed
Warning

A149558-100MG A149558-1G A149558-5G
77010-690EA 37 USD
77010-690 77010-692 77010-694
3-Amino-3-(3-methoxyphenyl)propan-1-ol 95%
3-Amino-3-(3-methoxyphenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MDL Number: MFCD03412652
CAS Number: 22490-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR