5-Chloro-1-methylisatin 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-1-methylindole-2,3-dione, 5-Chloro-1-methylindoline-2,3-dione, 5-Chloro-1-methyl-2,3-indolinedione

A149369-1G A149369-250MG A149369-5G
76984-574EA 63.66 USD
76984-574 76984-576 76984-578
5-Chloro-1-methylisatin 98%
5-Chloro-1-methylisatin

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MW: 195.60 g/mol
MDL Number: MFCD02956479
CAS Number: 60434-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR