(2R)-Morpholine-2-carboxylic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-Morpholine-2-carboxylic acid hydrochloride
A149304-1G
A149304-250MG
A149304-5G
77010-684EA
92.3
USD
77010-684
77010-686
77010-688
(2R)-Morpholine-2-carboxylic acid hydrochloride 97%
(2R)-Morpholine-2-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₉NO₃·ClH MW: 167.591 g/mol |
MDL Number:
MFCD13194711 CAS Number: 1273577-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...