Isoquinoline-7-carbonitrile 97%

Supplier: Ambeed
Warning

A149169-100MG A149169-1G A149169-250MG A149169-5G
77028-046EA 56.23 USD
77028-046 77028-048 77028-050 77028-052
Isoquinoline-7-carbonitrile 97%
Isoquinoline-7-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MW: 154.1680 g/mol
MDL Number: MFCD10699231
CAS Number: 223671-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR