Potassium 6-hydroxyhexanoate 97%

Supplier: Ambeed
Warning

A149061-100G A149061-1G A149061-250MG A149061-25G A149061-5G
77010-674EA 290 USD
77010-674 77010-676 77010-678 77010-680 77010-682
Potassium 6-hydroxyhexanoate 97%
Potassium 6-hydroxyhexanoate

Precaución: for Research Use Only

Formula: C₆H₁₁KO₃
MDL Number: MFCD27920725
CAS Number: 21810-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR