6-Chloro-4-methylpyridine-3-sulfonyl chloride 97%

Supplier: Ambeed
Danger

A149023-1G A149023-100MG A149023-250MG
76984-572EA 54 USD
76984-572 77889-980 77889-981
6-Chloro-4-methylpyridine-3-sulfonyl chloride 97%
6-Chloro-4-methylpyridine-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO₂S
MW: 226.08 g/mol
MDL Number: MFCD08436478
CAS Number: 889944-76-7
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR