1-Bromo-2,3-difluoro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

A148993-10G A148993-1G A148993-5G A148993-250MG A148993-25G
77028-036EA 285 USD
77028-036 77028-038 77028-040 77667-126 77667-128
1-Bromo-2,3-difluoro-4-nitrobenzene 98%
1-Bromo-2,3-difluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MDL Number: MFCD09839215
CAS Number: 1003708-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR