2-Ethylhexyl glycidyl ether 98%

Supplier: Ambeed
Warning

A148924-100G A148924-25G A148924-500G
76997-258EA 19 USD
76997-258 76997-260 76997-262
2-Ethylhexyl glycidyl ether 98%
2-Ethylhexyl glycidyl ether

Precaución: for Research Use Only

Formula: C₁₁H₂₂O₂
MDL Number: MFCD00005141
CAS Number: 2461-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR