Potassium trifluoro(2-(methylthio)pyrimidin-5-yl)borate 97%

Supplier: Ambeed
Warning

A148778-100MG A148778-1G A148778-250MG A148778-5G
76984-558EA 42 USD
76984-558 76984-560 76984-562 76984-564
Potassium trifluoro(2-(methylthio)pyrimidin-5-yl)borate 97%
Potassium trifluoro(2-(methylthio)pyrimidin-5-yl)borate

Precaución: for Research Use Only

Formula: C₅H₅BF₃KN₂S
MDL Number: MFCD18325143
CAS Number: 1245906-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR