Methyl 3-amino-3-(4-nitrophenyl)propanoate 95%

Supplier: Ambeed
Warning

A148726-250MG A148726-5G
76984-554EA 42 USD
76984-554 77889-976
Methyl 3-amino-3-(4-nitrophenyl)propanoate 95%
Methyl 3-amino-3-(4-nitrophenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₄
MDL Number: MFCD18907619
CAS Number: 273920-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR