3-Bromo-4,5-dimethylnitrobenzene 95%

Supplier: Ambeed
Warning

A148697-100MG A148697-250MG A148697-5G
77010-652EA 6 USD
77010-652 77010-654 77889-975
3-Bromo-4,5-dimethylnitrobenzene 95%
3-Bromo-4,5-dimethylnitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MDL Number: MFCD12547850
CAS Number: 22162-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR