2,7-Dichloro-alpha-[(dibutylamino)methyl]-9H-fluorene-4-methanol 97%

Supplier: Ambeed
Warning

A148677-100G A148677-10G A148677-25G
77028-006EA 91.24 USD
77028-006 77028-008 77028-010
2,7-Dichloro-alpha-[(dibutylamino)methyl]-9H-fluorene-4-methanol 97%
2,7-Dichloro-alpha-[(dibutylamino)methyl]-9H-fluorene-4-methanol

Precaución: for Research Use Only

Formula: C₂₃H₂₉Cl₂NO
MW: 406.39 g/mol
MDL Number: MFCD12923306
CAS Number: 69759-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR