Methyl-2-amino-5-bromothiazole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-5-bromothiazole-4-carboxylate

A148452-10G A148452-5G
77010-650EA 83.82 USD
77010-650 77592-800
Methyl-2-amino-5-bromothiazole-4-carboxylate 95%
Methyl-2-amino-5-bromothiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂S
MW: 237.08 g/mol
MDL Number: MFCD00137990
CAS Number: 850429-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR