Boc-ORN(Boc)-OH 97%

Supplier: Ambeed
Warning

A148414-1G A148414-25G A148414-5G A148414-10G
77027-988EA 5 USD
77027-988 77027-990 77027-992 77667-110
Boc-ORN(Boc)-OH 97%
Boc-ORN(Boc)-OH

Precaución: for Research Use Only

Formula: C₁₅H₂₈N₂O₆
MDL Number: MFCD00151882
CAS Number: 57133-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR