3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-phenylbutanoic acid 97%

Supplier: Ambeed
Warning

A148132-1G A148132-250MG
76997-226EA 132.62 USD
76997-226 76997-228
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-phenylbutanoic acid 97%
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄
MW: 401.45 g/mol
MDL Number: MFCD02090627
CAS Number: 282524-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR