3,4-Dimethyldiphenylamine 98%

Supplier: Ambeed
Warning

A147929-100G A147929-10G A147929-25G A147929-500G
76997-212EA 57 USD
76997-212 76997-214 76997-216 76997-218 76997-220
3,4-Dimethyldiphenylamine 98%
3,4-Dimethyldiphenylamine

Precaución: for Research Use Only

Formula: C14H15N
MDL Number: MFCD03093248
CAS Number: 17802-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR