2,2',7,7'-Tetrabromo-9,9'-spirobifluorene 98%

Supplier: Ambeed
Warning

A147830-10G A147830-1G A147830-25G A147830-5G A147830-100G
76997-198EA 53 USD
76997-198 76997-200 76997-202 76997-204 77549-816
2,2',7,7'-Tetrabromo-9,9'-spirobifluorene 98%
2,2',7,7'-Tetrabromo-9,9'-spirobifluorene

Precaución: for Research Use Only

Formula: C₂₅H₁₂Br₄
MDL Number: MFCD08704220
CAS Number: 128055-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR