2-Amino-6-methoxyphenol hydrochloride 98%

Supplier: Ambeed
Warning

A147787-100MG A147787-1G
76984-528EA 33 USD
76984-528 76984-530
2-Amino-6-methoxyphenol hydrochloride 98%
2-Amino-6-methoxyphenol hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀ClNO₂
MDL Number: MFCD24842777
CAS Number: 339531-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR